siRNA Details
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080776.1 | NP_542954.1 | 2 | 695 | ||
XM_006237921.2 | XP_006237983.1 | 2 | 761 | |||
GenBank mRNA | D63886.1 | - | 2 | 695 | ||
D85509.1 | - | 2 | 695 |
Gene Symbol: LOC498369 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770181.1 | XP_008768403.1 | 2 | 303 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019724, AF282844, AB021228, BC057926, AK053598, and others.
Gene Ontology Categories:
