siRNA Details
Gene Symbol: Gale | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080783.2 | NP_542961.2 | 4 | 289 | ||
XM_006239134.2 | XP_006239196.1 | 4 | 1254 | |||
XM_008764197.1 | XP_008762419.1 | 4 | 520 | |||
GenBank mRNA | BC097293.1 | - | 4 | 289 | 7443707 | |
X53949.1 | - | 3 | 270 |
