siRNA Details
Gene Symbol: Reln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080394.2 | NP_536319.2 | 13 | 1690 | ||
XM_006235873.2 | XP_006235935.1 | 13 | 1764 | |||
GenBank mRNA | AB049473.1 | - | 13 | 1858 | ||
AB062680.1 | - | 13 | 1690 |
