siRNA Details
Gene Symbol: Abcb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080582.1 | NP_542149.1 | 14 | 2044 | ||
GenBank mRNA | AF106563.1 | - | 6 | 431 | ||
AJ003004.1 | - | 14 | 2044 | |||
BC085712.1 | - | 14 | 2060 | 7190755 | ||
FQ211570.1 | - | 14 | 2070 |
