siRNA Details
Gene Symbol: Cacng3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080691.1 | NP_542422.1 | 1 | 130 | ||
GenBank mRNA | AF361340.1 | - | 1 | 130 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019430, BC145971, AJ272044, BC138673.
Gene Ontology Categories:
