siRNA Details
Gene Symbol: Chrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080773.1 | NP_542951.1 | 1 | 833 | ||
XM_006230974.2 | XP_006231036.1 | 2 | 1526 | |||
XM_008760215.1 | XP_008758437.1 | 2 | 1345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
