siRNA Details
Gene Symbol: P2rx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080780.2 | NP_542958.2 | 9, 10 | 1173 | ||
XM_006246566.2 | XP_006246628.1 | 8, 9 | 1181 | |||
GenBank mRNA | X92069.1 | - | 9, 10 | 1173 | ||
X97328.1 | - | 9, 10 | 1039 |
