siRNA Details
Gene Symbol: Dcbld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130419.1 | NP_569103.1 | 1, 2 | 215 | ||
GenBank mRNA | AF387549.1 | - | 1, 2 | 215 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028523, BC066097, AF387548.
Gene Ontology Categories:
