siRNA Details
Gene Symbol: Clec2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130402.2 | NP_569086.1 | 4 | 487 | ||
XM_008763309.1 | XP_008761531.1 | 5 | 984 | |||
GenBank mRNA | AF321552.1 | - | 6 | 673 | ||
EF100688.1 | - | 4 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
