siRNA Details
Gene Symbol: SULT1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014465.3 | NP_055280.2 | 3, 4 | 583 | ||
XM_005265676.3 | XP_005265733.1 | 4, 5 | 498 | |||
XM_005265677.3 | XP_005265734.1 | 4, 5 | 698 | |||
XM_005265678.3 | XP_005265735.1 | 3, 4 | 451 | |||
XM_011531873.2 | XP_011530175.1 | 2, 3 | 322 | |||
GenBank mRNA | BC010895.1 | - | 3, 4 | 585 | 4105139 | |
D89479.1 | - | 2, 3 | 309 | |||
U95726.1 | - | 2, 3 | 300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
