siRNA Details
Gene Symbol: Slc5a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130746.1 | NP_570102.1 | 18 | 2235 | ||
GenBank mRNA | AF026554.1 | - | 18 | 2235 | ||
AF081204.2 | - | 15 | 1823 | |||
AF081205.1 | - | 15 | 1823 | |||
BC168145.1 | - | 16 | 2097 | 8371761 | ||
FQ220013.1 | - | 4 | 332 |
