siRNA Details
Gene Symbol: Samsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130821.1 | NP_570834.1 | 6 | 571 | ||
GenBank mRNA | AB067679.1 | - | 6 | 571 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023380, DQ333189, BC052906, BC064778, AK041340, and others.
Gene Ontology Categories:
