siRNA Details
Gene Symbol: Mgea5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131904.1 | NP_571979.1 | 9 | 1367 | ||
GenBank mRNA | AY039679.1 | - | 9 | 1367 |

More Information
Other Species Targeted

Gene Ontology Categories:
