siRNA Details
Gene Symbol: Vom1r93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008971.1 | NP_001008971.1 | 1 | 408 | ||
XM_006236848.1 | XP_006236910.1 | 3 | 744 | |||
GenBank mRNA | AY510293.1 | - | 1 | 408 | ||
U36896.1 | - | 3 | 758 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
