siRNA Details
Gene Symbol: Zfp111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133323.1 | NP_579857.1 | 3 | 417 | ||
XM_006228375.2 | XP_006228437.1 | 1 | 357 | |||
XM_006228376.2 | XP_006228438.1 | 2 | 513 | |||
GenBank mRNA | FQ211918.1 | - | 3 | 483 | ||
U27186.1 | - | 3 | 417 |
Gene Symbol: LOC102548230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228505.2 | XP_006228567.1 | 2 | 1495 | ||
XM_006228506.2 | XP_006228568.1 | 3 | 751 | |||
XM_006228507.2 | XP_006228569.1 | 3 | 630 |
Gene Symbol: LOC102548503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759056.1 | XP_008757278.1 | 3 | 430 |
Gene Symbol: LOC102548600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759057.1 | XP_008757279.1 | 3 | 218 |
Gene Symbol: Zfp108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_574384.6 | XP_574384.4 | 1 | 199 |

More Information
Other Species Targeted

Gene Ontology Categories:
