siRNA Details
Gene Symbol: Tmlhe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133387.2 | NP_596878.2 | 5 | 627 | ||
XM_006255855.1 | XP_006255917.1 | 4 | 636 | |||
GenBank mRNA | AF374406.1 | - | 3 | 468 | ||
BC085923.1 | - | 5 | 627 | 7109209 |
