siRNA Details
Gene Symbol: Lonp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133404.1 | NP_596895.1 | 3, 4 | 601 | ||
GenBank mRNA | AB064323.1 | - | 3, 4 | 601 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lonp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133404.1 | NP_596895.1 | 3, 4 | 601 | ||
GenBank mRNA | AB064323.1 | - | 3, 4 | 601 |