siRNA Details
Gene Symbol: HGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004712.4 | NP_004703.1 | 5 | 521 | ||
XM_011525463.2 | XP_011523765.1 | 5 | 470 | |||
XM_017025297.1 | XP_016880786.1 | 6 | 678 | |||
XM_017025298.1 | XP_016880787.1 | 5 | 454 | |||
GenBank mRNA | AF260566.1 | - | 5 | 390 | ||
AK294190.1 | - | 5 | 469 | |||
AK303800.1 | - | 5 | 437 | |||
BC003565.1 | - | 5 | 463 | 3544197 | ||
BT009754.1 | - | 5 | 386 | |||
D84064.1 | - | 5 | 446 | |||
DR001213.1 | - | 5 | 521 | |||
KU178466.1 | - | 5 | 386 | |||
KU178467.1 | - | 5 | 386 | |||
U43895.1 | - | 5 | 461 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
