siRNA Details
Gene Symbol: LOC291863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013889.1 | NP_001013911.1 | 3 | 349 | ||
GenBank mRNA | BC078681.1 | - | 3 | 349 | 7123712 |
Gene Symbol: Ces1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133295.3 | NP_579829.3 | 3 | 374 | ||
GenBank mRNA | AF171640.1 | - | 3 | 323 | ||
BC061789.1 | - | 3 | 374 | 5623732 | ||
L46791.1 | - | 3 | 353 | |||
L81144.1 | - | 3 | 333 | |||
X51974.1 | - | 3 | 371 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
