siRNA Details
Gene Symbol: Dkc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133419.1 | NP_596910.1 | 14 | 1505 | ||
GenBank mRNA | BC087600.1 | - | 14 | 1464 | ||
BC099832.1 | - | 14 | 1463 | 7934021 | ||
Z34922.1 | - | 14 | 1505 |
Gene Symbol: LOC499971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024317.2 | NP_001019488.2 | 19 | 2484 | ||
XM_008762655.1 | XP_008760877.1 | 18 | 2137 | |||
XM_008762656.1 | XP_008760878.1 | 17 | 2028 | |||
GenBank mRNA | BC079392.1 | - | 12 | 1420 | 7111961 |
Gene Symbol: Smarca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134368.1 | NP_599195.1 | 10 | 2064 | ||
XM_006242596.2 | XP_006242658.1 | 10 | 2061 | |||
XM_006242597.2 | XP_006242659.1 | 10 | 2061 | |||
XM_006242598.2 | XP_006242660.1 | 10 | 2061 | |||
XM_006242599.2 | XP_006242661.1 | 10 | 2061 | |||
XM_006242600.2 | XP_006242662.1 | 10 | 2061 | |||
XM_006242601.2 | XP_006242663.1 | 10 | 2061 | |||
GenBank mRNA | AJ504723.1 | - | 9 | 1742 |

More Information
Other Species Targeted

Gene Ontology Categories:
