siRNA Details
Gene Symbol: Rab3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133536.2 | NP_598220.1 | 3 | 355 | ||
XM_008760719.1 | XP_008758941.1 | 3 | 520 | |||
GenBank mRNA | D78197.1 | - | 3 | 332 | ||
U54807.1 | - | 3 | 359 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023852, AB232589, AY026947, AF312037, BC114335, and others.
Gene Ontology Categories:
