siRNA Details
Gene Symbol: Sult1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133547.4 | NP_598231.3 | 4, 5 | 408 | ||
XM_006244242.2 | XP_006244304.1 | 3, 4 | 384 | |||
XM_006244243.2 | XP_006244305.1 | 3, 4 | 376 | |||
XM_006244245.2 | XP_006244307.1 | 3, 4 | 371 | |||
XM_006244246.2 | XP_006244308.1 | 3, 4 | 359 | |||
XM_006244248.2 | XP_006244310.1 | 4, 5 | 501 | |||
XM_006244249.2 | XP_006244311.1 | 2, 3 | 310 | |||
XM_006244250.2 | XP_006244312.1 | 2, 3 | 419 | |||
XM_008766738.1 | XP_008764960.1 | 3, 4 | 389 | |||
XM_008766739.1 | XP_008764961.1 | 3, 4 | 352 | |||
XM_008766740.1 | XP_008764962.1 | 3, 4 | 382 | |||
XM_008766741.1 | XP_008764963.1 | 3, 4 | 377 | |||
XM_008766742.1 | XP_008764964.1 | 3, 4 | 364 | |||
XM_008766743.1 | XP_008764965.1 | 3, 4 | 407 | |||
XM_008766744.1 | XP_008764966.1 | 2, 3 | 297 | |||
XM_008766745.1 | XP_008764967.1 | 2, 3 | 285 | |||
GenBank mRNA | AJ238391.1 | - | 1, 2 | 190 | ||
BC103636.1 | - | 4, 5 | 408 | 7106590 |
Gene Symbol: LOC100910057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244225.2 | XP_006244287.1 | 2, 3 | 336 | ||
XM_006244227.2 | XP_006244289.1 | 3, 4 | 373 | |||
XM_006244228.2 | XP_006244290.1 | 3, 4 | 678 | |||
XM_006244229.2 | XP_006244291.1 | 3, 4 | 368 | |||
XM_006244230.2 | XP_006244292.1 | 3, 4 | 673 | |||
XM_006244231.2 | XP_006244293.1 | 2, 3 | 314 | |||
XM_006244232.2 | XP_006244294.1 | 2, 3 | 294 | |||
XM_006244233.2 | XP_006244295.1 | 2, 3 | 599 | |||
XM_006244236.2 | XP_006244298.1 | 1 | 32 | |||
XM_008766781.1 | XP_008765003.1 | 3, 4 | 373 | |||
XM_008766782.1 | XP_008765004.1 | 3, 4 | 356 | |||
XM_008766783.1 | XP_008765005.1 | 3, 4 | 351 | |||
XM_008766784.1 | XP_008765006.1 | 3, 4 | 329 | |||
XM_008766785.1 | XP_008765007.1 | 3, 4 | 373 | |||
XM_008766786.1 | XP_008765008.1 | 3, 4 | 373 |
Gene Symbol: LOC100910526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244254.2 | XP_006244316.1 | 219 |
Gene Symbol: RGD1562392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750614.3 | XP_003750662.1 | 2, 3 | 317 | ||
XM_008766791.1 | XP_008765013.1 | 2, 3 | 296 |
Gene Symbol: Sult1c2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013177.2 | NP_001013195.2 | 309 | |||
GenBank mRNA | AJ238392.1 | - | 250 | |||
BC081691.1 | - | 206 | 7110136 | |||
BC092564.1 | - | 309 | 7372924 | |||
FQ209913.1 | - | 303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
