siRNA Details
Gene Symbol: Opa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133585.3 | NP_598269.3 | 21 | 2206 | ||
XM_006248497.1 | XP_006248559.1 | 23 | 2470 | |||
XM_006248498.1 | XP_006248560.1 | 22 | 2416 | |||
XM_006248499.1 | XP_006248561.1 | 22 | 2359 | |||
GenBank mRNA | AY333988.1 | - | 21 | 2115 | ||
AY510274.1 | - | 21 | 2261 | |||
BC111071.1 | - | 21 | 2206 | 7324017 | ||
U93197.1 | - | 5 | 412 |
