siRNA Details
Gene Symbol: Hadhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239786.1 | XP_006239848.1 | 4 | 663 | ||
XM_006239788.1 | XP_006239850.1 | 4 | 319 | |||
XM_006239789.2 | XP_006239851.1 | 4 | 594 | |||
GenBank mRNA | BC060545.1 | - | 4 | 246 | 6918791 | |
D16479.1 | - | 3 | 210 | |||
FM050540.1 | - | 4 | 274 |
