siRNA Details
Gene Symbol: Mgst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134349.3 | NP_599176.1 | 2, 3 | 192 | ||
GenBank mRNA | BC063150.1 | - | 2, 3 | 190 | 6922267 | |
BP494884.1 | - | 2, 3 | 191 | |||
CB711033.1 | - | 2, 3 | 208 | |||
FJ179404.1 | - | 1, 2 | 121 | |||
FQ209427.1 | - | 2, 3 | 213 | |||
FQ209464.1 | - | 2, 3 | 205 | |||
FQ209465.1 | - | 2, 3 | 212 | |||
FQ212841.1 | - | 2, 3 | 213 | |||
FQ218096.1 | - | 2, 3 | 195 | |||
FQ218830.1 | - | 2, 3 | 220 | |||
FQ219010.1 | - | 2, 3 | 214 | |||
FQ219410.1 | - | 2, 3 | 186 | |||
FQ219464.1 | - | 2, 3 | 218 | |||
FQ219581.1 | - | 2, 3 | 214 | |||
J03752.1 | - | 2, 3 | 176 |
