siRNA Details
Gene Symbol: Sult1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133547.4 | NP_598231.3 | 5, 6 | 534 | ||
XM_006244242.2 | XP_006244304.1 | 4, 5 | 510 | |||
XM_006244243.2 | XP_006244305.1 | 4, 5 | 502 | |||
XM_006244245.2 | XP_006244307.1 | 4, 5 | 497 | |||
XM_006244246.2 | XP_006244308.1 | 4, 5 | 485 | |||
XM_006244248.2 | XP_006244310.1 | 5, 6 | 627 | |||
XM_006244249.2 | XP_006244311.1 | 3, 4 | 436 | |||
XM_006244250.2 | XP_006244312.1 | 3, 4 | 545 | |||
XM_008766738.1 | XP_008764960.1 | 4, 5 | 515 | |||
XM_008766739.1 | XP_008764961.1 | 4, 5 | 478 | |||
XM_008766740.1 | XP_008764962.1 | 4, 5 | 508 | |||
XM_008766741.1 | XP_008764963.1 | 4, 5 | 503 | |||
XM_008766742.1 | XP_008764964.1 | 4, 5 | 490 | |||
XM_008766743.1 | XP_008764965.1 | 4, 5 | 533 | |||
XM_008766744.1 | XP_008764966.1 | 3, 4 | 423 | |||
XM_008766745.1 | XP_008764967.1 | 3, 4 | 411 | |||
GenBank mRNA | AJ238391.1 | - | 2, 3 | 316 | ||
BC103636.1 | - | 5, 6 | 534 | 7106590 |
Gene Symbol: LOC100910057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244225.2 | XP_006244287.1 | 3, 4 | 462 | ||
XM_006244227.2 | XP_006244289.1 | 4, 5 | 499 | |||
XM_006244228.2 | XP_006244290.1 | 4, 5 | 804 | |||
XM_006244229.2 | XP_006244291.1 | 4, 5 | 494 | |||
XM_006244230.2 | XP_006244292.1 | 4, 5 | 799 | |||
XM_006244231.2 | XP_006244293.1 | 3, 4 | 440 | |||
XM_006244232.2 | XP_006244294.1 | 3, 4 | 420 | |||
XM_006244233.2 | XP_006244295.1 | 3, 4 | 725 | |||
XM_006244236.2 | XP_006244298.1 | 1, 2 | 158 | |||
XM_008766781.1 | XP_008765003.1 | 4, 5 | 499 | |||
XM_008766782.1 | XP_008765004.1 | 4, 5 | 482 | |||
XM_008766783.1 | XP_008765005.1 | 4, 5 | 477 | |||
XM_008766784.1 | XP_008765006.1 | 4, 5 | 455 | |||
XM_008766785.1 | XP_008765007.1 | 4, 5 | 499 | |||
XM_008766786.1 | XP_008765008.1 | 4, 5 | 499 |
Gene Symbol: LOC100910526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244254.2 | XP_006244316.1 | 345 |
Gene Symbol: RGD1559960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244256.2 | XP_006244318.2 | 1 | 119 | ||
XM_008766792.1 | XP_008765014.1 | 1 | 119 |
Gene Symbol: RGD1562392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750614.3 | XP_003750662.1 | 3, 4 | 443 | ||
XM_008766791.1 | XP_008765013.1 | 3, 4 | 422 |
Gene Symbol: Sult1c2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013177.2 | NP_001013195.2 | 435 | |||
GenBank mRNA | AJ238392.1 | - | 376 | |||
BC081691.1 | - | 332 | 7110136 | |||
BC092564.1 | - | 435 | 7372924 | |||
FQ209913.1 | - | 429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
