siRNA Details
Gene Symbol: Sumo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133594.2 | NP_598278.1 | 2, 3 | 321 | ||
XM_008768397.1 | XP_008766619.1 | 3, 4 | 461 | |||
GenBank mRNA | BC058446.1 | - | 284 | 6921523 | ||
BC078746.1 | - | 2, 3 | 255 | 7125132 | ||
CF976332.1 | - | 325 | ||||
FQ221996.1 | - | 288 | ||||
FQ225379.1 | - | 263 | ||||
FQ225921.1 | - | 279 | ||||
FQ225989.1 | - | 279 | ||||
FQ230134.1 | - | 290 | ||||
FQ235035.1 | - | 2, 3 | 202 | |||
L79949.1 | - | 2, 3 | 268 |
Gene Symbol: Sumo2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729458.4 | XP_002729504.1 | 1 | 258 |

More Information
Other Species Targeted

Gene Ontology Categories:
