siRNA Details
Gene Symbol: Trpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136229.1 | NP_001129701.1 | 22, 23 | 3498 | ||
GenBank mRNA | AB040807.1 | - | 7, 8 | 1397 |

More Information
Other Species Targeted

Gene Ontology Categories:
