siRNA Details
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270589.1 | NP_001257518.1 | 3, 4 | 941 | ||
NM_001270590.1 | NP_001257519.1 | 3, 4 | 626 | |||
NM_001270591.1 | NP_001257520.1 | 4, 5 | 404 | |||
NM_001270592.1 | NP_001257521.1 | 4, 5 | 401 | |||
NM_001270593.1 | NP_001257522.1 | 3, 4 | 338 | |||
NM_001270594.1 | NP_001257523.1 | 3, 4 | 335 | |||
NM_134390.3 | NP_599217.1 | 3, 4 | 944 | |||
XM_006236455.2 | XP_006236517.1 | 3, 4 | 327 | |||
GenBank mRNA | AF370882.1 | - | 2, 3 | 297 | ||
BC059116.1 | - | 3, 4 | 443 | 6920949 | ||
FM070640.1 | - | 3, 4 | 325 | |||
FM086181.1 | - | 4, 5 | 388 | |||
FN804490.1 | - | 4, 5 | 387 | |||
FQ209634.1 | - | 3, 4 | 332 | |||
FQ212987.1 | - | 3, 4 | 333 | |||
FQ219149.1 | - | 4, 5 | 388 | |||
FQ219532.1 | - | 4, 5 | 388 | |||
FQ219576.1 | - | 3, 4 | 332 | |||
FQ220000.1 | - | 3, 4 | 517 | |||
FQ233370.1 | - | 4, 5 | 398 | |||
FQ235102.1 | - | 3, 4 | 350 |
