siRNA Details
Gene Symbol: Syne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227851.2 | XP_006227913.1 | 1354 | |||
XM_008758713.1 | XP_008756935.1 | 1366 | ||||
GenBank mRNA | AF452647.1 | - | 748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
