siRNA Details
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270589.1 | NP_001257518.1 | 6, 7 | 1351 | ||
NM_001270590.1 | NP_001257519.1 | 6, 7 | 1036 | |||
NM_001270591.1 | NP_001257520.1 | 7, 8 | 814 | |||
NM_001270592.1 | NP_001257521.1 | 7, 8 | 811 | |||
NM_001270593.1 | NP_001257522.1 | 6, 7 | 748 | |||
NM_001270594.1 | NP_001257523.1 | 6, 7 | 745 | |||
NM_134390.3 | NP_599217.1 | 6, 7 | 1354 | |||
XM_006236455.2 | XP_006236517.1 | 6, 7 | 737 | |||
GenBank mRNA | AF370882.1 | - | 5, 6 | 707 | ||
BC059116.1 | - | 6, 7 | 853 | 6920949 | ||
FQ209634.1 | - | 6, 7 | 742 | |||
FQ212987.1 | - | 6, 7 | 743 | |||
FQ219149.1 | - | 7, 8 | 798 | |||
FQ219532.1 | - | 7, 8 | 798 | |||
FQ219576.1 | - | 6, 7 | 742 | |||
FQ220000.1 | - | 6, 7 | 928 | |||
FQ233370.1 | - | 7, 8 | 808 | |||
FQ235102.1 | - | 6, 7 | 760 |
