siRNA Details
Gene Symbol: Stau2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007149.1 | NP_001007150.1 | 459 | |||
NM_001007150.1 | NP_001007151.1 | 186 | ||||
NM_134466.2 | NP_604461.2 | 282 | ||||
GenBank mRNA | AF483620.1 | - | 459 | |||
AF483621.1 | - | 459 | ||||
AY549445.1 | - | 186 | ||||
AY549446.1 | - | 186 | ||||
AY549447.1 | - | 282 | ||||
AY549448.1 | - | 282 | ||||
AY684789.1 | - | 186 | ||||
AY684790.1 | - | 186 | ||||
BC085705.1 | - | 818 | 7190916 |
