siRNA Details
Gene Symbol: LOC103690037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230507.2 | XP_006230569.2 | 3 | 3244 |
Gene Symbol: Ddx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135801.1 | NP_001129273.1 | 5 | 499 | ||
GenBank mRNA | BC160885.1 | - | 5 | 499 | 7384442 |
Gene Symbol: LOC100363112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177822.1 | NP_001171293.1 | 4 | 334 | ||
GenBank mRNA | AY325169.1 | - | 4 | 334 |
Gene Symbol: LOC499407 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047939.1 | NP_001041404.1 | 4 | 605 | ||
XM_006256227.2 | XP_006256289.2 | 4 | 4484 | |||
GenBank mRNA | AY387083.1 | - | 4 | 605 |
Gene Symbol: Nlrp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134375.3 | NP_599202.2 | 2080 | |||
GenBank mRNA | DQ631800.1 | - | 2080 | |||
M85183.1 | - | 956 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009946, NM_153065, NM_026186, NM_007929, NM_001004193, and others.
Gene Ontology Categories:
