siRNA Details
Gene Symbol: Svop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769315.1 | XP_008767537.1 | 9 | 1032 | ||
XM_008769316.1 | XP_008767538.1 | 9 | 1498 | |||
GenBank mRNA | AF060173.1 | - | 9 | 1127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
