siRNA Details
Gene Symbol: Exoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134414.2 | NP_599241.2 | 24, 25 | 2649 | ||
XM_006253891.1 | XP_006253953.1 | 25, 26 | 2721 | |||
XM_006253893.2 | XP_006253955.1 | 24, 25 | 2605 | |||
XM_006253895.2 | XP_006253957.1 | 23, 24 | 2533 | |||
GenBank mRNA | AF032666.1 | - | 24, 25 | 2587 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025588, BC049102, AK016532, BC048154, BC031207, and others.
Gene Ontology Categories:
