siRNA Details
Gene Symbol: Akr1c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138510.1 | NP_612519.1 | 5, 6 | 623 | ||
GenBank mRNA | BC086579.1 | - | 5, 6 | 622 | ||
BC100248.1 | - | 5, 6 | 617 | 7307214 | ||
D14424.1 | - | 5, 6 | 608 | |||
L32601.1 | - | 5, 6 | 623 |
Gene Symbol: Akr1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013057.1 | NP_001013075.1 | 6, 7 | 777 | ||
XM_006254183.2 | XP_006254245.1 | 6, 7 | 789 | |||
XM_006254184.2 | XP_006254246.1 | 6, 7 | 768 | |||
XM_006254185.2 | XP_006254247.1 | 5, 6 | 711 | |||
GenBank mRNA | BC078957.1 | - | 6, 7 | 777 | 7110806 |

More Information
Other Species Targeted

Gene Ontology Categories:
