siRNA Details
Gene Symbol: SULT4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014351.3 | NP_055166.1 | 4 | 615 | ||
GenBank mRNA | AF115311.1 | - | 4 | 499 | ||
AF176342.1 | - | 4 | 519 | |||
AF188698.1 | - | 4 | 582 | |||
AF251263.1 | - | 4 | 528 | |||
AK295399.1 | - | 4 | 448 | |||
AK313048.1 | - | 4 | 540 | |||
AL590119.1 | - | 4 | 499 | |||
BC022459.1 | - | 4 | 508 | 4795827 | ||
BC028171.1 | - | 4 | 587 | 5243984 | ||
BC030665.1 | - | 4 | 643 | 5258865 | ||
CR456588.1 | - | 4 | 500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
