siRNA Details
Gene Symbol: Dnajc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138856.2 | NP_620211.2 | 3, 4 | 315 | ||
XM_008760746.1 | XP_008758968.1 | 7, 8 | 1191 | |||
GenBank mRNA | AF492384.1 | - | 2, 3 | 238 | ||
BC086587.1 | - | 3, 4 | 315 | |||
BC158554.1 | - | 3, 4 | 534 | 7380100 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030046, BC048354, AK019636.
Gene Ontology Categories:
