siRNA Details
Gene Symbol: DCLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195415.1 | NP_001182344.1 | 14 | 4649 | ||
NM_001195416.1 | NP_001182345.1 | 13 | 4575 | |||
NM_004734.4 | NP_004725.1 | 18 | 5520 | |||
XM_005266592.2 | XP_005266649.1 | 17 | 5399 | |||
XM_006719893.2 | XP_006719956.1 | 17 | 5433 | |||
XM_017020847.1 | XP_016876336.1 | 17 | 5410 | |||
GenBank mRNA | AB002367.1 | - | 18 | 5449 | ||
BC152456.1 | - | 18 | 5449 | 8862589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
