siRNA Details
Gene Symbol: Oas1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138913.1 | NP_620268.1 | 2 | 329 | ||
XM_006249382.2 | XP_006249444.1 | 2 | 373 | |||
GenBank mRNA | AY221507.1 | - | 2 | 345 | ||
BC086979.1 | - | 2 | 316 | 7191865 | ||
FQ231421.1 | - | 2 | 358 | |||
FQ231433.1 | - | 2 | 334 | |||
FQ232784.1 | - | 2 | 359 | |||
Z18877.1 | - | 2 | 347 |
Gene Symbol: LOC100910735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751197.2 | XP_003751245.1 | 329 |
Gene Symbol: Oas1i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009680.1 | NP_001009680.1 | 2 | 309 | ||
GenBank mRNA | AY196697.1 | - | 2 | 309 | ||
BC160819.1 | - | 2 | 283 | 7382468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
