siRNA Details
Gene Symbol: Akr1c14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138547.2 | NP_612556.1 | 7, 8 | 896 | ||
GenBank mRNA | BC091123.1 | - | 7, 8 | 896 | 7372929 | |
D17310.1 | - | 7, 8 | 1084 | |||
M64393.1 | - | 7, 8 | 895 | |||
S57790.1 | - | 7, 8 | 854 |

More Information
Other Species Targeted

Gene Ontology Categories:
