siRNA Details
Gene Symbol: Akr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138884.1 | NP_620239.1 | 2 | 273 | ||
XM_006236285.1 | XP_006236347.1 | 2 | 293 | |||
GenBank mRNA | D17309.1 | - | 2 | 273 | ||
FQ211413.1 | - | 2 | 291 | |||
FQ219185.1 | - | 2 | 292 | |||
FQ219421.1 | - | 2 | 236 | |||
FQ219730.1 | - | 2 | 236 | |||
S80431.1 | - | 2 | 245 |
