siRNA Details
Gene Symbol: Ear11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138902.1 | NP_620257.1 | 409 | |||
GenBank mRNA | D88586.1 | - | 409 |
Gene Symbol: LOC100361866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728192.3 | XP_002728238.1 | 2 | 458 |
Gene Symbol: Rnase17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012232.1 | NP_001012232.1 | 1 | 346 | ||
XM_008770561.1 | XP_008768783.1 | 2 | 424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
