siRNA Details
Gene Symbol: Fnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138914.1 | NP_620269.1 | 188 | |||
XM_006233929.2 | XP_006233991.1 | 3 | 1584 | |||
GenBank mRNA | AB073208.1 | - | 188 | |||
AB126168.1 | - | 188 | ||||
AB126169.1 | - | 188 | ||||
AB126170.1 | - | 188 | ||||
AB126171.1 | - | 188 | ||||
AB126172.1 | - | 188 |
