siRNA Details
Gene Symbol: Ctsq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139262.1 | NP_640355.1 | 3, 4 | 298 | ||
GenBank mRNA | AF187323.1 | - | 3, 4 | 298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
