siRNA Details
Gene Symbol: Lrit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139331.2 | NP_647547.2 | 2 | 518 | ||
GenBank mRNA | AB028461.1 | - | 518 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139331.2 | NP_647547.2 | 2 | 518 | ||
GenBank mRNA | AB028461.1 | - | 518 |