siRNA Details
Gene Symbol: Uxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139336.1 | NP_647552.1 | 2, 3 | 176 | ||
XM_008766748.1 | XP_008764970.1 | 3, 4 | 677 | |||
XM_008766750.1 | XP_008764972.1 | 2, 3 | 221 | |||
XM_008766753.1 | XP_008764975.1 | 2, 3 | 569 | |||
GenBank mRNA | AF482705.1 | - | 2, 3 | 176 | ||
BC086988.1 | - | 2, 3 | 187 | 7192831 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026430, BC037049, AK005536, AK152376, AK075806, and others.
Gene Ontology Categories:
