siRNA Details
Gene Symbol: DCLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004734.4 | NP_004725.1 | 2 | 456 | ||
XM_005266592.2 | XP_005266649.1 | 2 | 409 | |||
XM_006719893.2 | XP_006719956.1 | 2 | 443 | |||
XM_017020847.1 | XP_016876336.1 | 2 | 409 | |||
XM_017020848.1 | XP_016876337.1 | 2 | 409 | |||
GenBank mRNA | AB002367.1 | - | 2 | 385 | ||
AK295102.1 | - | 2 | 409 | |||
BC152456.1 | - | 2 | 385 | 8862589 | ||
BQ711413.1 | - | 2 | 433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
