siRNA Details
Gene Symbol: Kcnc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139217.1 | NP_631963.1 | 3, 4 | 1819 | ||
XM_006241327.2 | XP_006241389.1 | 4, 5 | 2071 | |||
GenBank mRNA | M59211.1 | - | 3, 4 | 1819 | ||
M84203.1 | - | 3, 4 | 1862 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
