siRNA Details
Gene Symbol: RPS6KA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322227.1 | NP_001309156.1 | 18 | 3734 | ||
NM_001322228.1 | NP_001309157.1 | 16 | 3466 | |||
NM_001322229.1 | NP_001309158.1 | 17 | 3553 | |||
NM_001322230.1 | NP_001309159.1 | 18 | 3761 | |||
NM_001322231.1 | NP_001309160.1 | 16 | 3470 | |||
NM_001322232.1 | NP_001309161.1 | 16 | 3355 | |||
NM_001322233.1 | NP_001309162.1 | 16 | 3412 | |||
NM_001322234.1 | NP_001309163.1 | 17 | 3611 | |||
NM_001322235.1 | NP_001309164.1 | 18 | 3714 | |||
NM_001322236.1 | NP_001309165.1 | 16 | 3490 | |||
NM_001322237.1 | NP_001309166.1 | 17 | 3373 | |||
NM_001322238.1 | NP_001309167.1 | 15 | 3082 | |||
NM_004755.3 | NP_004746.2 | 17 | 3574 | |||
GenBank mRNA | AF090421.1 | - | 17 | 3522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
